872101C
Avanti
16:0-20:4 PE-N-20:4
Avanti Research™ - A Croda Brand 872101C
Synonym(s):
1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphoethanolamine-N-arachidonoyl (ammonium salt)
About This Item
Recommended Products
form
liquid
packaging
pkg of 1 × 2.5 mL (872101C-25mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 872101C
concentration
10 mg/mL (872101C-25mg)
lipid type
phosphoglycerides
bioactive lipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
[H][C@@](COP([O-])(OCCNC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+]
InChI
1S/C61H104NO9P.H3N/c1-4-7-10-13-16-19-22-25-27-29-31-34-36-39-42-45-48-51-59(63)62-54-55-69-72(66,67)70-57-58(56-68-60(64)52-49-46-43-40-37-33-24-21-18-15-12-9-6-3)71-61(65)53-50-47-44-41-38-35-32-30-28-26-23-20-17-14-11-8-5-2;/h16-17,19-20,25-28,31-32,34
InChI key
OTSYPQBHAMUVQA-HWEIRUDLSA-N
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3
Target Organs
Central nervous system
WGK
WGK 3
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service