Skip to Content
Merck
All Photos(1)

Key Documents

860544P

Avanti

C12 Galactosyl(β) Ceramide (d18:1/12:0)

Avanti Research - A Croda Brand 860544P, powder

Synonym(s):

D-galactosyl-β-1,1′ N-lauroyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H69NO8
CAS Number:
Molecular Weight:
643.93
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860544P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860544P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO)O

InChI

1S/C36H69NO8/c1-3-5-7-9-11-13-14-15-16-18-19-21-23-25-30(39)29(28-44-36-35(43)34(42)33(41)31(27-38)45-36)37-32(40)26-24-22-20-17-12-10-8-6-4-2/h23,25,29-31,33-36,38-39,41-43H,3-22,24,26-28H2,1-2H3,(H,37,40)/b25-23+/t29-,30+,31?,33-,34?,35?,36+/m0/s1

InChI key

IYCYEZLMOLRFAN-YJMGPQMXSA-N

General description

Galactosyl ceramide is a complex glycosphingolipid, which consists of a single galactose residue linked to ceramide. It is a major component of myelin sheath.

Application

C12 Galactosyl(β) Ceramide (d18:1/12:0) may be used as an internal standard in nerve lipidomics.

Biochem/physiol Actions

Galactosyl ceramide is involved in normal myelin function and stability.

Packaging

5 mL Amber Glass Screw Cap Vial (860544P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gangliosides in Health and Disease (2018)
Rebeca Jimeno et al.
eLife, 8 (2019-12-17)
Tissue homeostasis is critically dependent on the function of tissue-resident lymphocytes, including lipid-reactive invariant natural killer T (iNKT) cells. Yet, if and how the tissue environment shapes the antigen specificity of iNKT cells remains unknown. By analysing iNKT cells from
Pediatric Neurology (2013)
Monocarboxylate transporter 1 in Schwann cells contributes to maintenance of sensory nerve myelination during aging
Jha MK, et al.
Glia, 68(1), 161-177 (2020)
Divalent cation-mediated interaction between cerebroside sulfate and cerebrosides: an investigation of the effect of structural variations of lipids by electrospray ionization mass spectrometry
Koshy KM, et al.
Biophysical Journal, 77(1), 306-318 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service