Skip to Content
Merck
All Photos(2)

Key Documents

855677C

Avanti

17:1 Lyso PC

1-(10Z-heptadecenoyl)-2-hydroxy-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

17:1 LPC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H50NO7P
Molecular Weight:
507.64
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (855677C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855677C

concentration

10 mg/mL (855677C-10mg)

shipped in

dry ice

storage temp.

−20°C

Application

17:1 Lyso PC has been used as a lipid standard to optimize the mobile phase in the ultra-performance liquid chromatography.

Packaging

5 mL Clear Glass Sealed Ampule (855677C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of lipidomic platform and phosphatidylcholine retention time index for lipid profiling of rosuvastatin treated human plasma
Choi JM, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 944, 157-165 (2014)
Tsukasa Yagi et al.
Cancers, 12(1) (2020-01-01)
Ovarian cancer remains a highly lethal disease due to its late clinical presentation and lack of reliable early biomarkers. Protein-based diagnostic markers have presented limitations in identifying ovarian cancer. We tested the potential of phospholipids as markers of ovarian cancer
Jadegoud Yaligar et al.
PloS one, 9(3), e91436-e91436 (2014-03-19)
Dietary saturated fatty acids contribute to the development of fatty liver and have pathogenic link to systemic inflammation. We investigated the effects of dietary fat towards the pathogenesis of non-alcoholic fatty liver disease by longitudinal in vivo magnetic resonance spectroscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service