Skip to Content
Merck
All Photos(2)

Key Documents

850797C

Avanti

22:6 PE

1,2-didocosahexaenoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

1,2-di-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phosphoethanolamine; PE(22:6(4Z,7Z,10Z,13Z,16Z,19Z)/22:6(4Z,7Z,10Z,13Z,16Z,19Z)) ; 110931

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H74NO8P
CAS Number:
Molecular Weight:
836.09
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850797C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850797C

concentration

10 mg/mL (850797C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O)COC(CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O

InChI

1S/C49H74NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-41-48(51)55-45-47(46-57-59(53,54)56-44-43-50)58-49(52)42-40-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h5-8,11-14,17-20,23-26,29-32,35-38,47H,3-4,9-10,15-16,21-22,27-28,33-34,39-46,50H2,1-2H3,(H,53,54)/b7-5-,8-6-,13-11-,14-12-,19-17-,20-18-,25-23-,26-24-,31-29-,32-30-,37-35-,38-36-/t47-/m1/s1

InChI key

RBDNSJMXMWTPCS-NYUVKXDYSA-N

General description

22:6 PE is a polyunsaturated homoacid. Phosphatidylethanolamine (PE) is a membrane phospholipid. The types of PE depends on the combination of free fatty acids on the α and β position. 22:6 PE denotes the length of the carbon chain:degree of unsaturation in the PE.

Application

22:6 PE has been used as a standard for the optimization of liquid chromatography-mass spectrometry (LC/MS) conditions and to detect the serum antiphosphatidylethanolamine (aPE) antibodies from patients using enzyme linked immunosorbent assay (ELISA). It is suitable for use in testing the effect of increasing acyl chain unsaturation on phase separation from the lipid raft molecules spingomyelin (SM) and cholesterol (CHOL).

Biochem/physiol Actions

Phosphatidylethanolamine (PE) plays a key role in membrane fusion, cell cycle, autophagy, apoptosis, and cognitive function.

Packaging

5 mL Clear Glass Sealed Ampule (850797C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acyl chain unsaturation in PEs modulates phase separation from lipid raft molecules
Shaikh SR, et al.
Biochemical and Biophysical Research Communications, 311(3), 793-796 (2003)
The outcome of ELISA for antiphosphatidylethanolamine antibodies is dependent on the composition of phosphatidylethanolamine
Hou S, et al.
Journal of Immunological Methods, 440, 27-34 (2017)
Diarachidonoylphosphoethanolamine induces necrosis/necroptosis of malignant pleural mesothelioma cells
Kaku Y, et al.
Cellular Signalling, 27(9), 1713-1719 (2015)
Comprehensive untargeted lipidomic analysis using core-shell C30 particle column and high field orbitrap mass spectrometer
Narvaez-Rivas M and Zhang Q
Journal of Chromatography A, 1440, 123-134 (2016)
Mónica Narváez-Rivas et al.
Journal of chromatography. A, 1440, 123-134 (2016-03-02)
The goal of untargeted lipidomics is to have high throughput, yet comprehensive and unambiguous identification and quantification of lipids. Novel stationary phases in LC separation and new mass spectrometric instruments capable of high mass resolving power and faster scanning rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service