Skip to Content
Merck
All Photos(2)

Key Documents

850454P

Avanti

16:0-14:0 PC

1-palmitoyl-2-myristoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-hexadecanoyl-2-tetradecanoyl-sn-glycero-3-phosphocholine; PMPC; PC(16:0/14:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H76NO8P
CAS Number:
Molecular Weight:
705.99
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850454P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850454P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI key

UIXXHROAQSBBOV-PSXMRANNSA-N

General description

Phosphatidylcholine (PC) is an important component of the mucosal layer of the colon. 16:0-14:0 PC (PMPC/1-palmitoyl-2-myristoyl-sn-glycero-3-phosphocholine) is the positional isomer of 1-myristoyl-2-palmitoyl-PC (MPPC).

Application

16:0-14:0 PC (1-palmitoyl-2-myristoyl-sn-glycero-3-phosphocholine) may be used as an inhibitor for screening several recombinant human UDP-glucuronosyltransferases (UGTs).

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance.

Packaging

5 mL Amber Glass Screw Cap Vial (850454P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Xin Gao et al.
Xenobiotica; the fate of foreign compounds in biological systems, 45(3), 197-206 (2014-09-27)
1.Endogenous compounds have been reported to be the regulators of UDP-glucuronosyltransferases (UGTs) isoforms. This study aims to investigate the regulatory effects of the activity of UGT isoforms by two important lipid components phosphatidylcholine (PC) and lysophosphatidylcholines (LPC) using in vitro
Integrative Medicine - E-Book (2017)
Hitoshi Matsuki et al.
International journal of molecular sciences, 14(2), 2282-2302 (2013-01-26)
Bilayers formed by phospholipids are frequently used as model biological membranes in various life science studies. A characteristic feature of phospholipid bilayers is to undergo a structural change called a phase transition in response to environmental changes of their surroundings.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service