Skip to Content
Merck
All Photos(2)

Key Documents

840862C

Avanti

18:0-18:2 PA

1-stearoyl-2-linoleoyl-sn-glycero-3-phosphate (sodium salt), chloroform

Synonym(s):

1-octadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phosphate (sodium salt); PA(18:0/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H72O8PNa
CAS Number:
Molecular Weight:
722.95
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840862C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840862C

concentration

10 mg/mL (840862C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCCCC)O

InChI

1S/C39H73O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(40)45-35-37(36-46-48(42,43)44)47-39(41)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h12,14,18,20,37H,3-11,13,15-17,19,21-36H2,1-2H3,(H2,42,43,44);/q;+1/p-1/b14-12-,20-18-;/t37-;/m1./s1

InChI key

QCCKEVYCMGRONT-ZNKFQGFOSA-M

General description

Phosphatidic acid (PA) is an amphipathic lipid, containing a polar head group and two long hydrophobic acyl chains. PA is anionic in nature and a membrane structural lipid. It acts as a parent compound for more complex membrane phospholipids.

Biochem/physiol Actions

Phosphatidic acid (PA) is a bioactive lipid and a secondary messenger. PA reacts with water and other phospholipids to produce lipid bilayers and multicomponent bilayers respectively.

Packaging

5 mL Clear Glass Sealed Ampule (840862C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioactive Lipids
An Introduction to Biological Membranes, 453-478 (2016)
Lipid second messengers and Receptors
Encyclopedia of Endocrine Diseases, 77(3), 329-334 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service