Skip to Content
Merck
All Photos(2)

Key Documents

840038P

Avanti

12:0 PS

1,2-dilauroyl-sn-glycero-3-phospho-L-serine (sodium salt), powder

Synonym(s):

1,2-didodecanoyl-sn-glycero-3-phospho-L-serine (sodium salt); DLPS; PS(12:0/12:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H57NO10PNa
CAS Number:
Molecular Weight:
645.74
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (840038P-10mg)
pkg of 1 × 200 mg (840038P-200mg)
pkg of 1 × 25 mg (840038P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840038P

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCCCCCCCCCC)=O)COC(CCCCCCCCCCC)=O.[Na+]

InChI

1S/C30H58NO10P.Na/c1-3-5-7-9-11-13-15-17-19-21-28(32)38-23-26(24-39-42(36,37)40-25-27(31)30(34)35)41-29(33)22-20-18-16-14-12-10-8-6-4-2;/h26-27H,3-25,31H2,1-2H3,(H,34,35)(H,36,37);/q;+1/p-1/t26-,27+;/m1./s1

InChI key

QBSSGICBQYAHPS-OUPRKWGVSA-M

General description

Phosphatidylserine (PS), an aminophospholipid is negatively charged and accounts for 2–10% of total cellular lipid. It is located in the membrane leaflets.

Application

12:0 PS may be used:
  • in the preparation of large unilamellar vesicles (LUVs)
  • as a standard to spike replicas of extracted lipid droplets and calibration curve generation for lipidomic analysis
  • to study its effects on α-secretase activity

Biochem/physiol Actions

Phosphatidylserine (PS) plays a key role in protein kinase C pathways. It also helps in confining the intracellular proteins to membrane leaflets of cytosol.

Packaging

5 mL Amber Glass Screw Cap Vial (840038P-10mg)
5 mL Amber Glass Screw Cap Vial (840038P-200mg)
5 mL Amber Glass Screw Cap Vial (840038P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristof Schutters et al.
Apoptosis : an international journal on programmed cell death, 15(9), 1072-1082 (2010-05-05)
Cells are able to execute apoptosis by activating series of specific biochemical reactions. One of the most prominent characteristics of cell death is the externalization of phosphatidylserine (PS), which in healthy cells resides predominantly in the inner leaflet of the
Bao Fu Qiao et al.
The journal of physical chemistry. B, 117(17), 5073-5080 (2013-04-10)
Crystalline vesicles are promising nanomaterials due to their mechanical stability in various environments. To control their fabrication, it is essential to understand the effects of different experimental conditions on crystallization. Here we perform atomistic molecular dynamics simulations of anionic lipid
C Preston Moon et al.
Journal of molecular biology, 413(2), 484-494 (2011-09-06)
The free energy of unfolding of a membrane protein from lipids into water (ΔG(o)(w,l)) describes its equilibrium thermodynamic stability. Knowing this parameter gives insight into a membrane protein's sequence-structure-energy relationships. However, there are few measures of membrane protein stability because
Marcus O W Grimm et al.
International journal of molecular sciences, 14(3), 5879-5898 (2013-03-15)
Alzheimer's disease (AD) is characterized by extracellular accumulation of amyloid-β peptide (Aβ), generated by proteolytic processing of the amyloid precursor protein (APP) by β- and γ-secretase. Aβ generation is inhibited when the initial ectodomain shedding is caused by α-secretase, cleaving
Alexander Fauland et al.
Journal of lipid research, 52(12), 2314-2322 (2011-10-01)
This work aims to combine chromatographic retention, high mass resolution and accuracy, MS/MS spectra, and a package for automated identification and quantitation of lipid species in one platform for lipidomic analysis. The instrumental setup elaborated comprises reversed-phase HPLC coupled to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service