Skip to Content
Merck
All Photos(1)

Key Documents

780201C

Avanti

18:1 PE MCC

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide] (sodium salt), chloroform

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide] (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H90N2NaO11P
CAS Number:
Molecular Weight:
985.25
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (780201C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 780201C

concentration

10 mg/mL (780201C-25mg)

shipped in

dry ice

storage temp.

−20°C

General description

18:1 PE MCC (1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide]) lipid comprising of phosphoethanolamine linked to two oleic acid via its phosphate group and to a maleimide group via its amino group. It is a maleimide-functionalized thiol-reactive lipid.

Application

18:1 PE MCC 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide] has been used:
  • in liposome preparation for cysteine coupling studies
  • in nanodisc impregnation for ras sarcoma protein (K-RAS4B) tethering
  • in liposome preparation for single molecule imaging

Packaging

5 mL Clear Glass Sealed Ampule (780201C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liposome-based assays to study membrane-associated protein networks
Methods in Enzymology, 534, 223-243 (2014)
Walter Huynh et al.
The Journal of cell biology, 216(10), 3051-3060 (2017-09-09)
Bicaudal D2 (BICD2) joins dynein with dynactin into a ternary complex (termed DDB) capable of processive movement. Point mutations in the BICD2 gene have been identified in patients with a dominant form of spinal muscular atrophy, but how these mutations
Zhenhao Fang et al.
Cell chemical biology, 25(11), 1327-1336 (2018-08-21)
KRAS is frequently mutated in several of the most lethal types of cancer; however, the KRAS protein has proven a challenging drug target. K-RAS4B must be localized to the plasma membrane by prenylation to activate oncogenic signaling, thus we endeavored
Jenny J Lin et al.
Biophysical journal, 118(12), 2879-2893 (2020-05-15)
The natural peptide-major histocompatibility complex (pMHC) ligand for T cell receptors (TCRs) is inactive from solution yet capable of activating T cells at single-molecule levels when membrane-associated. This distinctive feature stems from the mechanism of TCR activation, which is thought to involve

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service