Skip to Content
Merck
All Photos(1)

Key Documents

330500

Avanti

Inositol Snoopers

Avanti Research - A Croda Brand 330500

Synonym(s):

Inositol Snoopers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.62

manufacturer/tradename

Avanti Research - A Croda Brand 330500

packaging

1 ea of 10 × (membrane packaged in foil pouch (330500-10EA))
1 ea of 5 × (membrane packaged in foil pouch (330500-5EA))

shipped in

wet ice

storage temp.

2-8°C

General description

Avanti Inositol Snoopers consist of 13 different lipid species spotted individually on a solid support. Each spot contains 1 μg of the highest quality pure lipid and is ideal for investigating lipid-protein interactions.

Application

Inositol Snoopers has been used to obtain qualitative information about the binding of phosphatase and tensin homolog (pten) and its derivatives to anionic lipids.It has also been used to study lipid-protein interactions, characterize lipid-binding antibodies and identify novel lipid-interacting proteins of potential therapeutic value.

Packaging

Membrane Packaged in Foil Pouch (330500-10EA)
Membrane Packaged in Foil Pouch (330500-5EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Snoopers is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biophysical methods for the characterization of PTEN/lipid bilayer interactions
Harishchandra RK, et al.
Methods, 77, 125-135 (2015)
Oriol Gallego et al.
Molecular systems biology, 6, 430-430 (2010-12-02)
Protein-metabolite networks are central to biological systems, but are incompletely understood. Here, we report a screen to catalog protein-lipid interactions in yeast. We used arrays of 56 metabolites to measure lipid-binding fingerprints of 172 proteins, including 91 with predicted lipid-binding
Ronald C Bruntz et al.
The Journal of biological chemistry, 289(2), 600-616 (2013-11-22)
The lack of innovative drug targets for glioblastoma multiforme (GBM) limits patient survival to approximately 1 year following diagnosis. The pro-survival kinase Akt provides an ideal target for the treatment of GBM as Akt signaling is frequently activated in this
Avanti lipid tools: connecting lipids, technology, and cell biology
Sims KH, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1841(8), 1038-1048 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service