Skip to Content
Merck
All Photos(1)

Key Documents

300112S

Avanti

Dox-NP (2mL vial)

Avanti Research - A Croda Brand

Synonym(s):

Dox-NP (2mL vial)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

description

Dox-NP (Liposomal Encapsulated Doxorubicin)
frozen liposomal suspension (buffer - 10% w/w sucrose, 10 mM histidine ~pH 6.5)

Assay

>99% (HPLC/MS)

form

dispersion

packaging

pkg of 1 × 2 mL (300112S-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

shipped in

dry ice

storage temp.

−20°C

Application

Dox-NP(2mL vial) is suitable for use:
  • in ultraviolet-visible (UV-vis) spectra, cryo-transmission electron microscopy (cryo-TEM) and analytical ultracentrifuge experiments
  • in transmission electron microscopy (TEM) imaging studies
  • to analyse the effect of gold nanoparticle loaded liposomes on anti-tumor and cytotoxicity properties of DOX-NP on Ehrlich tumor

Biochem/physiol Actions

Dox-NP (doxorubicin loaded nanoparticles) is a liposomal drug encapsulating doxorubicin. This encapsulation reduces the side effects of doxorubicin and increases the effective local dose.

Packaging

10 mL amber serum vial (300112S-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Dox-NP is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transmission electron microscopy as a tool for the characterization of soft materials: application and interpretation
Franken LE, et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 4(5), 1600476-1600476 (2017)
Analytical ultracentrifugation for analysis of doxorubicin loaded liposomes
Mehn D, et al.
International Journal of Pharmaceutics, 523(1), 320-326 (2017)
Enhanced Ehrlich tumor inhibition using DOX-NP and gold nanoparticles loaded liposomes
Mady MM, et al.
Pakistan Journal of Pharmaceutical Sciences, 1(28), 2321-2325 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service