Skip to Content
Merck
All Photos(1)

Key Documents

190000C

Avanti

Yeast Extract Total

Avanti Research - A Croda Brand

Synonym(s):

Yeast Extract Total, Yeast Total Lipid Extract (S. cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

form

chloroform solution

packaging

pkg of 1 × 4 mL (190000C-100mg)
pkg of 5 × 4 mL (190000C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

25 mg/mL (190000C-100mg)
25 mg/mL (190000C-500mg)

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Total Lipid Extract is a chloroform:methanol extract of the respective tissue (Brain, Soybean, Heart, etc.). This extract is partitioned against deionized water, and the chloroform phase is concentrated.

Application

Yeast Extract Total is suitable as a reference standard for visualization and separation of the wide range of lipid classes present in biological samples.

Packaging

5 mL Clear Glass Sealed Ampule (190000C-100mg)
5 mL Clear Glass Sealed Ampule (190000C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Klose et al.
The Journal of biological chemistry, 285(39), 30224-30232 (2010-07-22)
The lipid raft concept proposes that biological membranes have the potential to form functional domains based on a selective interaction between sphingolipids and sterols. These domains seem to be involved in signal transduction and vesicular sorting of proteins and lipids.
Metabolomics and lipidomics using traveling-wave ion mobility mass spectrometry.
Paglia G and Astarita G
Nature Protocols, 12(4), 797-797 (2017)
Filip Duša et al.
Biochimica et biophysica acta. Biomembranes, 1862(2), 183115-183115 (2019-11-11)
The cell membrane is mainly composed of lipid bilayers with inserted proteins and carbohydrates. Lipid bilayers made of purified or synthetic lipids are widely used for estimating the effect of target compounds on cell membranes. However, the composition of such
Giuseppe Paglia et al.
Nature protocols, 12(4), 797-813 (2017-03-17)
Metabolomics and lipidomics aim to profile the wide range of metabolites and lipids that are present in biological samples. Recently, ion mobility spectrometry (IMS) has been used to support metabolomics and lipidomics applications to facilitate the separation and the identification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service