Documents
Z516481
Sigma-Aldrich
Aldrich® 5 mm NMR tube evacuation manifold
2 ports, L 150 mm
Synonym(s):
nmr tube, nmr tubes, tube manifold
About This Item
UNSPSC Code:
41121800
NACRES:
SB.32
L
150 mm
General description
Designed to evacuate NMR tubes that have a J Young PTFE valve. Once the NMR tube has been evacuated in the manifold, close the valve and place into the NMR spectrometer. Manifolds have 0-10 mm bore PTFE valves. The multiple evacuation station units allow removal of one NMR tube while the remaining tube(s) stay under vacuum.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC