Key Documents
Z220094
Sigma-Aldrich
Aldrich® short-path distillation head
Joints: ST/NS 24/40 (2), thermometer joint: ST/NS 10/30
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Joint
thermometer joint: ST/NS10/30
Joints: ST/NS24/40 (2)
immersion depth
75 mm
Related Categories
General description
Vigreux indents prevent bumping of fine or lightweight column packing into the condenser or fraction cutter when column is flooded.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)