Z202835
Solvent still heads
capacity 250 mL, bottom joint: ST/NS 24/40
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Joint
bottom joint: ST/NS24/40
side joint ST/NS14/20
top joint ST/NS24/40
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
General description
With return finger and 4 mm PTFE stopcock.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service