Skip to Content
Merck
All Photos(1)

Key Documents

Z177377

Sigma-Aldrich

Scienceware® micro funnel

top I.D. 43 mm, stem diam. 8 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.43

packaging

pack of 12 ea

manufacturer/tradename

Bel-Art F14685-0043

stem diam.

8 mm

top I.D.

43 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

PP. Useful for micro work, filling burettes, manometers, etc.

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service