Z154881
Disposable polystyrene weighing dishes
L × W × H 86 mm × 86 mm × 25 mm, white
Synonym(s):
Polystyrene Weighing Dishes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.71
Recommended Products
material
polystyrene
rectangular
packaging
pack of 100 ea
case of 500 ea
manufacturer/tradename
Sigma-Aldrich
L × W × H
86 mm × 86 mm × 25 mm
dish L × W
86 mm × 86 mm
color
white
Looking for similar products? Visit Product Comparison Guide
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hanna Tay et al.
Cells, 10(1) (2020-12-31)
Macrophages play an important but poorly understood role in angiogenesis. To investigate their role in vessel formation, relevant in vivo models are crucial. Although the chick chorioallantoic membrane (CAM) model has been frequently used as an angiogenesis assay, limited data
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service