Key Documents
Z122890
Ampule breaker/collar
For 5-10 mL ampules
About This Item
UNSPSC Code:
41121807
NACRES:
NB.43
Pricing and availability is not currently available.
General description
How to Use:
1. Slip polyethylene breaker/ collar over top of ampule.
2. Grip bottom of ampule with one hand and breaker/collar with other.
3. Gently snap top of ampule off.
4. Discard collar and ampule top.
Choose from one of the most recent versions:
Certificates of Analysis (COA)