Key Documents
Z101133
Stainless steel 304 syringe needle
gauge 20, L 12 in.
Synonym(s):
Stainless Needle, Steel Syringe
About This Item
UNSPSC Code:
42142600
eCl@ss:
32070500
NACRES:
NB.03
Pricing and availability is not currently available.
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 1 ea
manufacturer/tradename
Cadence Science 4188
L
12 in.
gauge
20
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Please, I'd to like to know the point style of this needle. Thank You!
    1 answer
    1. This is a deflected point Plated Brass Hub.
      Helpful?