WHAWB100032
Qiagen® swabs for bucal and saliva sample collection
foam swab, sterile, pkg of 100 ea
Synonym(s):
Foam Swab, Sterile (100), Whatman FTA Cards, Z747386
About This Item
Recommended Products
sterility
sterile
packaging
pkg of 100 ea
manufacturer/tradename
Qiagen WB100032
application(s)
environmental
food and beverages
suitability
bacteria
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
This article describes the evaluation of Whatman FTA cards from Cytiva for their ability to collect, store, and isolate high-quality RNA from a variety of crude biological samples.
Protocols
This study examined the yield and quality of DNA from samples applied to Whatman FTA cards, using five common methods of DNA extraction.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service