WHA10445830
Whatman® multiple vacuum filtration device
3-place, AS300/3, stainless steel, capacity 500 mL, 47/50, support screen
Synonym(s):
Z751057
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
stainless steel
stainless steel housing
feature
47/50, support screen
packaging
pk of 1
manufacturer/tradename
Whatman 10445830
Whatman Article No. 28415610 (US reference)
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- The stainless steel manifold is fitted for three or six filtration units.
- The apparatus can be autoclaved and sterilized by dry heat at up to 180°C.*Suitable only for vacuum operation.
- If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure)
- Applications includes: Microbiology quality control, residue analyses, serial filtration carried out rapidly and easily with a common drainage outlet.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service