Skip to Content
Merck
All Photos(1)

Key Documents

W478955

Sigma-Aldrich

2,6-Dimethyl-5-heptenol

natural, ≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18O
CAS Number:
Molecular Weight:
142.24
FEMA Number:
4789
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

botanical

Quality Level

grade

Halal
Kosher
natural

Agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 117

Assay

≥95%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; oily; waxy

InChI

1S/C9H18O/c1-8(2)5-4-6-9(3)7-10/h5,9-10H,4,6-7H2,1-3H3

InChI key

WFZFXUZFKAOTRR-UHFFFAOYSA-N

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

386.1 °F

Flash Point(C)

196.7 °F


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service