Skip to Content
Merck
All Photos(1)

Key Documents

W373923

Sigma-Aldrich

4-Vinylphenol solution

10 wt. % in propylene glycol, FG

Synonym(s):

4-ethenylphenol, p-Hydroxystyrene, p-vinylphenol

Sign Into View Organizational & Contract Pricing


About This Item

FEMA Number:
3739
Council of Europe no.:
11257
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.057
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

concentration

10 wt. % in propylene glycol

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

medicinal; chemical; phenolic; sweet

InChI

1S/C8H8O/c1-2-7-3-5-8(9)6-4-7/h2-6,9H,1H2

InChI key

FUGYGGDSWSUORM-UHFFFAOYSA-N

General description

4-Vinylphenol solution has been used as an HPLC internal standard during the identification of 4-vinyl derivatives formed via decarboxylation of hydroxycinnamic acids extracted from corn cobs using phenolic acid decarboxylase (PAD) enzyme extracted from Lactobacillus plantarum.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 2 - STOT RE 1

Target Organs

Stomach,intestinal tract,Kidney,Bladder

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioproduction of 4-vinylphenol from corn cob alkaline hydrolyzate in two-phase extractive fermentation using free or immobilized recombinant E. coli expressing pad gene.
Salgado JM, et al.
Enzyme and Microbial Technology, 58, 22-28 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service