Skip to Content
Merck
All Photos(1)

Key Documents

W347418

Sigma-Aldrich

2,6,6-Trimethyl-1-cyclohexene-1-acetaldehyde

≥75%

Synonym(s):

β-Homocyclocitral

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18O
CAS Number:
Molecular Weight:
166.26
FEMA Number:
3474
EC Number:
Council of Europe no.:
10338
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.112
NACRES:
NA.21
grade:
Fragrance grade
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Fragrance grade
Halal

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥75%

impurities

≤20% β-cyclocitral

refractive index

n20/D 1.485 (lit.)

bp

58-59 °C/0.4 mmHg (lit.)

density

0.941 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

oily; woody; cooling; camphoraceous; fruity

storage temp.

2-8°C

SMILES string

[H]C(=O)CC1=C(C)CCCC1(C)C

InChI

1S/C11H18O/c1-9-5-4-7-11(2,3)10(9)6-8-12/h8H,4-7H2,1-3H3

InChI key

VHTFHZGAMYUZEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service