Skip to Content
Merck
All Photos(1)

Key Documents

W270008

Sigma-Aldrich

Methyl cyclopentenolone

anhydrous, 98%, FCC, FG

Synonym(s):

Cyclotene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
FEMA Number:
2700
EC Number:
Council of Europe no.:
758
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.056
NACRES:
NA.21
Organoleptic:
caramel; coffee; maple; roasted; sweet
grade:
FG
Fragrance grade
Halal
Kosher
anhydrous
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
anhydrous

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

98%

mp

104-108 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

caramel; coffee; maple; roasted; sweet

SMILES string

CC1CCC(=O)C1=O

InChI

1S/C6H8O2/c1-4-2-3-5(7)6(4)8/h8H,2-3H2,1H3

InChI key

CFAKWWQIUFSQFU-UHFFFAOYSA-N

Related Categories

General description

Methyl cyclopentenolone has been identified in maple syrup[1], Wasanbon sugar[2] and raw cane sugar.[3]

Application

This material is found primarily as the enol form (CAS 80-71-7) in its pure, anhydrous state. Readily forms the diketone form (CAS 765-70-8) in solution.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F

Flash Point(C)

100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on the ?sugary flavor? of raw cane sugar.
Abe E, et al.
Proceedings of the Japan Academy. Series B, Physical and Biological Sciences, 54(9), 542-547 (1978)
The identification of methylcyclopentenolone and other compounds in maple sirup flavor extract.
Filipic VJ, et al.
Journal of Food Science, 30(6), 1008-1015 (1965)
Contents and Compositions of the Aroma in ?Wasanbon? Sugar.
Matsui T & Kitaoka S.
J. Nutr. Sci. Vitaminol., 27(6), 563-572 (1981)
Seo Young Choi et al.
Biochimica et biophysica acta, 1770(12), 1612-1619 (2007-10-13)
Peroxisome proliferator-activated receptors (PPARs) are members of the nuclear hormone receptor superfamily that plays a pivotal role in regulating inflammatory gene expression. The purpose of this study was to investigate the effects of coffee extract, 3-methyl-1,2-cyclopentanedione (3-MCP) on PPARs in
Bryson W Katona et al.
The Journal of organic chemistry, 72(24), 9298-9307 (2007-10-26)
Deoxycholic acid (DCA) is an endogenous secondary bile acid implicated in numerous pathological conditions including colon cancer formation and progression and cholestatic liver disease. DCA involvement in these disease processes results partly from its ability to modulate signaling cascades within

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service