Skip to Content
Merck
All Photos(2)

Key Documents

T5206

Sigma-Aldrich

Tetrabromophthalic anhydride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8Br4O3
CAS Number:
Molecular Weight:
463.70
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.11 mmHg ( 180 °C)

Assay

98%

mp

269-271 °C (lit.)

SMILES string

Brc1c(Br)c(Br)c2C(=O)OC(=O)c2c1Br

InChI

1S/C8Br4O3/c9-3-1-2(8(14)15-7(1)13)4(10)6(12)5(3)11

InChI key

QHWKHLYUUZGSCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Rahman et al.
The Science of the total environment, 275(1-3), 1-17 (2001-08-03)
Polybrominated diphenyl ether, PBDE, flame retardants are now a world-wide pollution problem reaching even remote areas. They have been found to bioaccumulate and there are concerns over the health effects of exposure to PBDEs, they also have potential endocrine disrupting
A Scozzafava et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 10(1), 29-41 (2000-03-04)
N-1-Tritylsulfenyl histamine was synthesized by reaction of histamine (Hst) with tetrabromophthalic anhydride followed by protection of its imidazole moiety with tritylsulfenyl chloride. After hydrazinolysis, it afforded a key intermediate which was derivatized at the aminoethyl group in order to obtain
Amelie Tsoutsoulopoulos et al.
Toxicology in vitro : an international journal published in association with BIBRA, 58, 245-255 (2019-03-21)
The CULTEX® Radial Flow System (RFS) is a modular in vitro system for the homogenous exposure of cells to airborne particles at the air-liquid interface (ALI). A former pre-validation study successfully demonstrated the general applicability of the CULTEX® RFS and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service