Key Documents
SYNR7010
Synthware high vacuum valve plug
size 0-4 mm
Synonym(s):
Synthware R7010
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware R7010
Related Categories
General description
Complete with control knob and PTFE stem having three O-rings.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)