Key Documents
SYNM150004
Synthware manifold with high vacuum stopcocks and #15 o-ring joints
Ports: 4
Synonym(s):
Synthware M150004, vacuum manifold, vacuum unit
About This Item
UNSPSC Code:
40161501
NACRES:
NB.77
manufacturer/tradename
M150004
General description
Synthware glass manifold featuring 4 mm double hollow high vacuum stopcocks, this four-port manifold is fitted with two 14/20 and two 24/40 inner joints, with a 100 mm center-to-center distance between the joints. The #15 O-ring joint stopper (SYNS480015) and the #15 O-ring joint to serrated hose adapter must be ordered separately.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)