Skip to Content
Merck
All Photos(1)

Key Documents

SYNF394100

Synthware powder funnel with 60 degree offset

joint: ST/NS 24/40, top diam. 100 mm

Synonym(s):

Synthware F394100, glass powder funnel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.51

Joint

joint: ST/NS24/40

packaging

pkg of 1 ea

manufacturer/tradename

Synthware F394100

top diam.

100 mm

Looking for similar products? Visit Product Comparison Guide

General description

For use with multi-neck flasks.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service