Skip to Content
Merck
All Photos(1)

Key Documents

QBD11162

Sigma-Aldrich

DOTA-tris(acid)-amido-dPEG ®12-TFP ester

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H83F10N5O25
Molecular Weight:
1380.23
UNSPSC Code:
12352106
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chelator
reagent type: cross-linking reagent
reagent type: fluorescent-labelling reagent

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

DOTA-tris(acid)-amido-dPEG®₁₂-TFP ester contains a DOTA-tris(acid) metal chelator for radio-labelling connected through a hydrophilic dPEG spacer to a TFP ester which can react with a primary amine.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service