Skip to Content
Merck
All Photos(1)

Key Documents

N21200

Sigma-Aldrich

2-Nitro-1,4-phenylenediamine

95%

Synonym(s):

1,4-Diamino-2-nitrobenzene, 2-Nitro-p-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(NH2)2
CAS Number:
Molecular Weight:
153.14
Beilstein:
2210195
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

135-138 °C (lit.)

SMILES string

Nc1ccc(N)c(c1)[N+]([O-])=O

InChI

1S/C6H7N3O2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H,7-8H2

InChI key

HVHNMNGARPCGGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>372.2 °F - Pensky-Martens closed cup

Flash Point(C)

> 189 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

I S Grover et al.
Mutation research, 300(1), 1-3 (1993-06-01)
The water and chloroform extracts of guava were tested for their antimutagenicity. The water extract was effective in inactivating the mutagenicity of direct-acting mutagens, e.g., 4-nitro-o-phenylenediamine, sodium azide, and the S9-dependent mutagen, 2-aminofluorene, in the tester strains of Salmonella typhimurium.
1,4-Diamino-2-nitrobenzene (2-nitro-para-phenylenediamine).
IARC monographs on the evaluation of carcinogenic risks to humans, 57, 185-200 (1993-01-01)
Metabolism of the hair dye component, nitro-p-phenylenediamine, in the rat.
M Nakao et al.
Chemical & pharmaceutical bulletin, 35(2), 785-791 (1987-02-01)
S Bala et al.
Mutation research, 222(3), 141-148 (1989-03-01)
The antimutagenic effect of 10 citrus fruit juices was observed against the mutagenicity of N-nitro-o-phenylenediamine (NPD) in TA97a and sodium azide in TA100 tester strains of Salmonella typhimurium using the Ames test. It was noticed that the juices of all
In vivo skin penetration studies of 2,4-toluenediamine, 2,4-diaminoanisole, 2-nitro-p-phenylenediamine, p-dioxane and N-nitrosodiethanolamine in cosmetics.
F N Marzulli et al.
Food and cosmetics toxicology, 19(6), 743-747 (1981-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service