MT51350008
Mettler-Toledo technical buffer
pH 9.21, volume 250 mL
About This Item
Recommended Products
sterility
non-sterile
form
liquid
manufacturer/tradename
Mettler-Toledo 51350008
volume
250 mL
pH
9.21
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- High-quality Mettler-Toledo Rainbow I buffers for accurate sensor calibration and maintenance.
- Small volume packaging (single-use sachets) ensures guaranteed fresh solutions and ease of use.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH SVHC Candidate List
EU REACH Annex XVII (Restriction List)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service