Skip to Content
Merck
All Photos(1)

Key Documents

L050080

Sigma-Aldrich

Cap B

configured for PerkinElmer, configured for Polygen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.21

type

for DNA synthesis

product line

Proligo Reagents

form

liquid

storage condition

dry at room temperature

impurities

≤200 ppm water content (Karl Fischer)

color

clear

suitability

conforms to structure (GC, Formulation)

compatibility

configured for PerkinElmer
configured for Polygen

Looking for similar products? Visit Product Comparison Guide

General description

Cap B solution contains tetrahydrofuran (THF), pyridine, and N-methylimidazole (NMI). It is mixed in situ with Cap A solution during the capping (acetylation) reaction. Pyridine is applied as a mild base while NMI provides a powerful acylation catalyst.

Preparation Note

Tetrahydrofurane / N-Methylimidazole / Pyridine 8/1/1 (v/v/v)

Other Notes

Cap B solution, containing tetrahydrofuran (THF), pyridine and N-methylimidazole (NMI), is mixed in situ with Cap A solution during the capping (acetylation) reaction. Pyridine is applied as a mild base while NMI provides a powerful acylation catalyst.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

6.1 °F

Flash Point(C)

-14.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service