material
gold/chromium electrode (Au/Cr)
description
die layer thickness 300 nm SiO2
die layer thickness 36 nm boron nitride
electrode layer thickness 60 nm gold
feature
BN/SiO2 coating
optimized for hall
packaging
pkg of 16 non-diced devices
manufacturer/tradename
HF101
die thickness
525 μm , silicon
electrode size
10 nm , chrome
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service