GF93887386
Polyamide - Nylon 6, 6
(PA 6,6), granule, 3 mm nominal granule size, weight 1000 g, grade super-tough
Synonym(s):
AM326315
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
granular
manufacturer/tradename
Goodfellow 938-873-86
weight
1000 g
nominal granule size
3 mm
Related Categories
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service