GF38090099
Niobium-zirconium alloy, Nb99Zr1
1000mm tube, 3.95mm outside diameter, 0.35mm wall thickness, 3.25mm inside diameter, stress relieved
Synonym(s):
NB037350
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
tubes
manufacturer/tradename
Goodfellow 380-900-99
L × wall thickness
1000 mm × 0.35 mm
O.D. × I.D.
3.95 mm × 3.25 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service