GF36604964
Quartz - fused
tube, outside diameter 4.0 mm, length 100 mm, wall thickness 1.0 mm
Synonym(s):
SI617050
About This Item
Recommended Products
form
tubes
manufacturer/tradename
Goodfellow 366-049-64
L
100 mm
O.D.
4.0 mm
wall thickness
1.0 mm
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service