GF14525209
Silver
foil, 6mm disks, thickness 0.125mm, annealed, 99.95+%
Synonym(s):
Silver, AG000401
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23
Assay
99.95%
form
foil
manufacturer/tradename
Goodfellow 145-252-09
resistivity
1.59 μΩ-cm, 20°C
diam. × thickness
6 mm × 0.125 mm
bp
2212 °C (lit.)
mp
960 °C (lit.)
density
10.49 g/cm3 (lit.)
SMILES string
[Ag]
InChI
1S/Ag
InChI key
BQCADISMDOOEFD-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Eugene A Chu et al.
Otology & neurotology : official publication of the American Otological Society, American Neurotology Society [and] European Academy of Otology and Neurotology, 24(3), 507-518 (2003-06-14)
To present the rich and checkered history of the artificial eardrum, a widely used device in the 19th century, and to illustrate the behavior of otologists in response to the introduction of a promising new technology. Over 40 published books
G J Hoffman et al.
Science (New York, N.Y.), 177(4051), 802-804 (1972-09-01)
The water potential of leaves in situ can be measured without temperature control with a miniature, single-junction psychrometer constructed from silver foil and attached to the leaf with a silver-impregnated, conductive coating. The temperature of the psychrometer has been found
D C Sharma et al.
Indian journal of physiology and pharmacology, 41(3), 285-288 (1997-07-01)
About 50 mg of silver leaf (metallic silver) was given daily by mouth to 30 healthy volunteers for 20 days. A statistically significant hypophospholipidemic, hypotriglyceridemic, hypocholesterolemic and hypoglycemic effect was observed. This was accompanied by a less marked fall in
IX. Note on Silver Foil in Surgery.
J S Lewis
Annals of surgery, 50(4), 793-796 (1909-10-01)
M Dubiel et al.
Journal of synchrotron radiation, 8(Pt 2), 539-541 (2001-08-22)
The thermal expansion behaviour of silver fcc has been described by an anharmonic Einstein model using EXAFS data in the temperature range between 10 and 300 K. The linear expansion coefficient of a bulk silver foil agrees well with X-ray
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service