Skip to Content
Merck
All Photos(1)

Key Documents

DWKWGF541P

Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture

label, Ethanol, low-density polyethylene bottle, capacity 500 mL

Synonym(s):

WGF541P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

material

low-density polyethylene bottle

feature

label Ethanol

packaging

pkg of 5 ea

manufacturer/tradename

Azlon WGF541P

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for ethanol dispensing within the laboratory

Features and Benefits

Slim design for comfortable handling and ease of use

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service