DWKCPS0500P
Azlon Squat Form Cylinder with Molded Graduation with blue over print
polypropylene cylinder, class B, capacity 500 mL
Synonym(s):
CPS0500P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
polypropylene cylinder
feature
class B
graduated
packaging
pkg of 1 ea
manufacturer/tradename
Azlon CPS0500P
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Offers increased stability especially when working in confined spaces
Features and Benefits
Unique self draining base to eliminate water collection during cleaning in laboratory washing machines
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service