BMSH2012
Benchmark Incu-Shaker™ 10LR Refrigerated Shaking Incubator with nonslip rubber mat
AC/DC input 115 V
Synonym(s):
shaker, incubator, incubated shaker, magic clamp, culture
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
Benchmark Scientific H2012
parameter
temp. range at Ambient -15 to 60 °C
30-300 rpm speed
W × L × H
45 cm × 45 cm × 40 cm, Internal dimensions
plate W × L
45 cm × 45 cm
Looking for similar products? Visit Product Comparison Guide
General description
Incu-Shaker™ 10LR is a heavyduty performer, designed forheavy workloads and continuous use. Its outstanding temperature accuracy anduniformity are supported by proprietary heat distribution technology thatensures that temperatures are precisely maintained throughout the chamber.Inconsistencies due to stratification are reduced to negligible levels and the selectedtemperature is precisely maintained within 0.25%, even in the corners. Theexpanded temperature range, down to 15 degrees below ambient, allows for abroad range of applications.
Application
Incubated shaking of various samples
Legal Information
Incu-Shaker is a trademark of Benchmark Scientific, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service