BAH369460000
SP Bel-Art® Sterileware™ Long Handled Sampling Spoon
white polystyrene, nominal capacity 14.79 mL , 3tsp, L × W 17.8 cm (7 in.) × 1.5625 in. (40 mm) mL, sterile, pack of 200 ea (individually wrapped)
Synonym(s):
Lab Scoop, dispensing tool, spoon
All Photos(4)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Recommended Products
material
white polystyrene
sterility
sterile
packaging
pack of 200 ea (individually wrapped)
manufacturer/tradename
Bel-Art H36946-0000
L × W
17.8 cm (7 in.) × 1.5625 in. (40 mm) mL
nominal capacity
14.79 mL , 3tsp
Related Categories
General description
Designed to gather small amounts of sample material, especially in hard to reach containers.
Features and Benefits
- Long handle reaches into jars, bottles and other confined areas
- Minimizes the potential of personal contact with sampled material
- Sterile, FDA grade polystyrene safe for food, drug and cosmetic use
- Individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing
Note: Sterilized products are not returnable.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)