BAH369441010
SP Bel-Art® Sterileware™ Long Handled Sampling Spoon
white polystyrene, nominal capacity 4.93 mL , 1tsp, L × W 17.8 cm (7 in.) × 1.1875 in. (30 mm), sterile, pack of 10 ea (double wrapped)
Synonym(s):
Lab Scoop, dispensing tool, spoon
All Photos(3)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
Recommended Products
material
white polystyrene
sterility
sterile
packaging
pack of 10 ea (double wrapped)
manufacturer/tradename
Bel-Art H36944-1010
L × W
17.8 cm (7 in.) × 1.1875 in. (30 mm)
nominal capacity
4.93 mL , 1tsp
Related Categories
General description
The long handle on these spoons enables users to reach into jars, bottles and other confined areas to minimize the potential of personal contact with sampled material.
Features and Benefits
- Ten individually packaged spoons are sealed in an outer bag to create multiples layers of protection against contaminants and debris
- Ideal for use in multi-zone clean rooms
- FDA grade polystyrene safe for food, drug and cosmetic use
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)