Skip to Content
Merck
All Photos(2)

Documents

B402

Sigma-Aldrich

Batyl alcohol

99%

Synonym(s):

1-O-Octadecyl-rac-glycerol, rac-Glycerol 1-octadecyl ether, DL-3-Octadecyloxy-1,2-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17OCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
344.57
Beilstein:
1725677
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

71-73 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCOCC(O)CO

InChI

1S/C21H44O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-24-20-21(23)19-22/h21-23H,2-20H2,1H3

InChI key

OGBUMNBNEWYMNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Feng Sun et al.
Biotechnology letters, 27(2), 113-117 (2005-02-11)
Novel mutual pro-drugs in which 2',3'-dideoxyinosine anti-HIV (human immunodeficiency virus) drug and 3-octadecyloxy-propane-1,2-diol anti-inflammatory drug were attached to the same molecule via different biodegradable linkages, were synthesized through two-step enzymatic transesterification by a commercial lipase in acetone.
A K Das et al.
Lipids, 27(6), 401-405 (1992-06-01)
Chronic feeding of 1-O-octadecyl-sn-glycerol (batyl alcohol) to patients suffering from congenital deficiency in tissue ether glycerolipids showed an increase in the plasmalogens content of their erythrocytes. However, nothing is known about the ether lipid content of other tissues in these
Treatment of peripheral leukopenia after cadaveric kidney transplantation.
B Xiwen et al.
Transplantation proceedings, 28(3), 1631-1632 (1996-06-01)
D Meusel et al.
Chemistry and physics of lipids, 61(2), 193-198 (1992-04-01)
Stereoselectivity of several triacylglycerol lipases (EC 3.1.1.3) has been investigated in the enzymatic esterification of rac-1-O-octadecylglycerol with oleic acid in the presence of organic solvents, such as hexane. X-1(3)-O-Octadecylmonooleoylglycerols were the only products formed with most lipases; considerable proportions of
N Yamamoto et al.
Cancer research, 48(21), 6044-6049 (1988-11-01)
Alkylglycerols, inflammation products of lipids in cancerous tissues, are potent macrophage stimulating agents. Administration of small amounts (10-100 ng) of alkylglycerols to mice greatly enhanced macrophage activation for Fc-mediated ingestion activity at the 5th day posttreatment. Dose effect analysis revealed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service