Skip to Content
Merck
All Photos(2)

Documents

932744

Sigma-Aldrich

Tris[2-(p-tolyl)pyridine]iridium(III)

≥99% (HPLC)

Synonym(s):

Ir(p-Me-ppy)3 , Ir(mppy)3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H30IrN3
CAS Number:
Molecular Weight:
696.86
UNSPSC Code:
12352302
NACRES:
NA.21

grade

sublimed grade

Quality Level

Assay

≥99% (HPLC)

loss

0.5% TGA, >330°C (weight loss)

solubility

chloroform: soluble
dichloromethane: soluble
toluene: soluble

λmax

287 nm±5 nm in dichloromethane
373 nm±5 nm in dichloromethane

fluorescence

λem 514 nm±10 nm in dichloromethane

Orbital energy

HOMO 5.6 eV 
LUMO 3.0 eV 

Looking for similar products? Visit Product Comparison Guide

Application

Tris[2-(p-tolyl)pyridine]iridium(III), also known as Ir(mppy)3, is a solution-processable small molecule, used as green phosphorescent dopant in highly-efficient light emitting diode (LED) displays, both perovskite and organic LEDs.
Tris[2-(p-tolyl)pyridine]iridium(III) is widely utilized as a phosphorescent emitter in OLED devices, particularly in green or yellow organic light-emitting diodes (OLEDs). It can be used in R&D studies for the development of advanced display technologies, including flat-panel displays and organic electroluminescent (OEL) displays. It is suitable in studies such as solid-state lighting including white OLEDs (WOLEDs) and organic light-emitting transistors (OLETs). It has been studied as a sensitizer in dye-sensitized solar cells (DSSCs), as a component in hybrid perovskite solar cells. It can be used in the research area of chemical sensor applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient green single-emitting layer phosphorescent organic light-emitting diodes with an iridium(III) complex as a hole-type sensitizer
Wu R, et al.
Journal of Material Chemistry C, 7, 2744-2750 (2019)
Organic Light-Emitting Diodes with 30% External Quantum Efficiency Based on a Horizontally Oriented Emitter
Kim, Sei-Yong, et al.
Advanced Functional Materials , 23, 3896-3900 (2013)
Kebin Lin et al.
Nature, 562(7726), 245-248 (2018-10-12)
Metal halide perovskite materials are an emerging class of solution-processable semiconductors with considerable potential for use in optoelectronic devices1-3. For example, light-emitting diodes (LEDs) based on these materials could see application in flat-panel displays and solid-state lighting, owing to their
Xin Ai et al.
Nature, 563(7732), 536-540 (2018-11-23)
Organic light-emitting diodes (OLEDs)1-5, quantum-dot-based LEDs6-10, perovskite-based LEDs11-13 and micro-LEDs14,15 have been championed to fabricate lightweight and flexible units for next-generation displays and active lighting. Although there are already some high-end commercial products based on OLEDs, costs must decrease whilst maintaining high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service