Skip to Content
Merck
All Photos(2)

Key Documents

900704

Sigma-Aldrich

Graphene oxide

greener alternative

paste, non-exfoliated

Synonym(s):

Graphene oxide nanoplatelets

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CxHyOz
UNSPSC Code:
12352119
NACRES:
NA.23

Quality Level

form

paste

composition

C, ≥3% (by gravimetric analysis combined with EA)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

General description

Handling of large quantities of GO powder is not recommended since graphene oxide powder is thermally unstable. Graphene oxide paste can make storage and transportation of large quantities in a safe manner. Graphene oxide solutions of desired concentration or graphene oxide paper or sheet can be made from graphene oxide paste. Sonication is required to obtain single layers of graphene oxide. To make a concentrated aqueous dispersion that will not settle under gravity: add 25 mL water/1 g paste, stir overnight for 16 h, sonicate for 30 minutes in a mild bath sonicator. Centrifuge (effective diameter of the rotor is 23 cm) at 3500 rpm for 10 minutes to remove any large unexfoliated particles and decant the supernate to give ∼2 mg/mL graphene oxide in water. The above concentrate can be diluted and sonicated further to get single layer graphene oxide.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Graphene based inks are ideal elements for 3D printed energy storage devices. Click here for more information.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". High concentrated graphene oxide sheets provide the prerequisite viscosity to bind the electrode materials together and enable 3D printing. Using water as a green solvent makes this aqueous ink system feasible for processing and drying safety and low cost. Click here for more information.

Application

  • Composites.
  • Inks and coatings.
  • Transparent conductive films.
  • Supercapacitors.
  • Batteries.
  • Graphene oxide sheets at interfaces.
This product can be used in material extrusion 3D printing technique.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Seungae Lee et al.
Nanoscale, 8(40), 17551-17559 (2016-10-08)
A highly transparent paste adhesive is successfully fabricated by introducing graphene oxide (GO) to silicone paste adhesive by using a solvent-exchange method. The GO incorporated in the paste adhesive has a significant role in improving thermal conductivity, transparency and adhesive
Amperometric enzyme-free glucose sensor based on the use of a reduced graphene oxide paste electrode modified with electrodeposited cobalt oxide nanoparticles.
Hassan H, and Esmaeil H
Microchimica Acta, 183(7), 2259-2266 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service