Skip to Content
Merck
All Photos(1)

Key Documents

900556

Sigma-Aldrich

Rare earth doped upconversion nanoparticles

fluorescence λem 540 nm (main peak), 10 mg/mL in toluene

Synonym(s):

CANdot® Series X up green, Er/NaYF4, Luminescent nanoparticles, NaYF4:Yb, Upconversion nanocrystals

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352303
NACRES:
NA.23

form

dispersion
nanoparticles

Quality Level

composition

Er/NaYF4
NaYF4:Yb

concentration

10 mg/mL in toluene

particle size

20 nm

λmax

980 nm

fluorescence

λem 540 nm (main peak)
λem 540 nm±15 nm

functional group

oleic acid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Our rare earth doped upconversion nanoparticles possess inorganic fluoride host lattices and can be easily excited with near-infra-red (NIR: 980 nm) light source. These Cd-free nanoparticles exhibit characteristic emission peaks and are well suited for applications as luminescent biological labels and as spectral converters for increasing solar cell efficiency.

Legal Information

Fraunhofer CAN is a research division of the Fraunhofer IAP
CANdot is a registered trademark of Fraunhofer CAN

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

43.0 °F

Flash Point(C)

6.11 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced luminescence efficiency of aqueous dispersible NaYF4:Yb/Er nanoparticles and the effect of surface coating.
Ansari AA, et al.
Royal Society of Chemistry Advances, 6, 22074-22082 (2016)
Cytotoxic effects of upconversion nanoparticles in primary hippocampal cultures.
Vedunova MV, et al.
Royal Society of Chemistry Advances, 6, 33656-33665 (2016)
Thorben Rinkel et al.
Angewandte Chemie (International ed. in English), 55(3), 1164-1167 (2015-12-04)
A new method is presented for preparing gram amounts of very small core/shell upconversion nanocrystals without additional codoping of the particles. First, ca. 5 nm β-NaYF4:Yb,Er core particles are formed by the reaction of sodium oleate, rare-earth oleate, and ammonium fluoride

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service