Skip to Content
Merck
All Photos(1)

Key Documents

797871

Sigma-Aldrich

Ultrastable fluorescent silica nanobeads

120 nm, lyophilized

Synonym(s):

Fluorescent beads, Fluorescent nanoparticles, Silica nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
Molecular Weight:
60.08
UNSPSC Code:
12352302
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

form

powder

particle size

120 nm

fluorescence

λem 590 nm

UV absorption

λ: 570 nm Amax

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

The use of labeling or staining agents has greatly assisted the study of complex biological interactions in biology and the fluorescent labeling of biomolecules has been demonstrated as an indispensable tool in many biological studies. By virtue of excellent fluorescence properties the silica nanobeads find application in biological labeling, bioimaging, biosensing etc.[1][2] Ultrastable particles exhibit a very high resistance to photobleaching and can be used in single-particle imaging as well as in super resolution techniques such as stimulated emission depletion (STED) microscopy.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luminescent nanomaterials for biological labelling
F. Wang, W. B. Tan, Y. Zhang, et al.
Nanotechnology, 17(1), R1-R13 (2006)
Geometrical confinement of quantum dots in porous nanobeads with ultraefficient fluorescence for cell-specific targeting and bioimaging
P-J Chen, S-H Hu, W-T Hung, S-Y Chen, and D-M Liu,
Journal of Materials Chemistry, 22(19), 9568-9575 (2012)

Articles

RAFT polymerization controls chain growth, yields defined polymers without cytotoxic heavy metals like ATRP.

RAFT polymerization controls chain growth, yields defined polymers without cytotoxic heavy metals like ATRP.

RAFT polymerization controls chain growth, yields defined polymers without cytotoxic heavy metals like ATRP.

RAFT polymerization controls chain growth, yields defined polymers without cytotoxic heavy metals like ATRP.

See All

Questions

  1. I want to know what is on the surface of this particle. Can I get azide functional group on the surface?

    1 answer
    1. Product 797871 is Ultrastable fluorescent silica nanobeads. There are OH groups on the surface. Fluorescent dye with similar properties to Atto 565 is used. It will be a custom product if azide needs to be grown on the surface.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service