Skip to Content
Merck
All Photos(2)

Key Documents

769037

Sigma-Aldrich

Propane

99.95%, Messer® CANGas

Synonym(s):

n-Propane, Propane gas

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH3
CAS Number:
Molecular Weight:
44.10
Beilstein:
1730718
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.5 (vs air)

Quality Level

vapor pressure

190 psi ( 37.7 °C)
8.42 atm ( 21.1 °C)

description

Filling mass: 15g
Filling volume: 7L

Assay

99.95%

form

gas

autoignition temp.

842 °F

expl. lim.

9.5 %

bp

-42.1 °C (lit.)

mp

-188 °C (lit.)

density

0.564 g/mL at 20 °C (lit.)

SMILES string

CCC

InChI

1S/C3H8/c1-3-2/h3H2,1-2H3

InChI key

ATUOYWHBWRKTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Propane is a colourless, odourless gas that is flammable, liquefied, and heavier than air. It is used as fuel for laboratory burners, portable stoves, and vehicles (liquefied petroleum gas [LPG]). Propane is also used in the research of polymerization catalysts. Gas is easy to handle in a small, light pressure can, while the disposable container allows for aluminium recycling when finished.

Recommended products

Z742374: CANgas pressure regulator
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve

Legal Information

Messer is a registered trademark of Messer Group GmbH

accessory

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-155.2 °F - closed cup

Flash Point(C)

-104 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Maksim N Khannanov et al.
Applied spectroscopy, 74(12), 1496-1504 (2020-03-13)
The low accessibility of natural gas fields and transporting pipelines requires portable online analyzers of the composition of natural gas, ensuring nearly chromatographic precision and capable of in situ analysis of a wide range of gases, including infrared-inactive ones (hydrogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service