Skip to Content
Merck
All Photos(1)

Key Documents

729841

Sigma-Aldrich

Triethylene glycol methyl ether methacrylate

contains MEHQ as inhibitor, 93%

Synonym(s):

Methoxy TEG methacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O5
CAS Number:
Molecular Weight:
232.27
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

93%

form

liquid

mol wt

average Mn 200

contains

MEHQ as inhibitor
1500-2500 ppm MEHQ as inhibitor

refractive index

n20/D 1.446

density

1.027 g/mL at 25 °C

Ω-end

methacrylate

α-end

methoxy

storage temp.

2-8°C

SMILES string

COCCOCCOCCOC(=O)C(C)=C

InChI

1S/C11H20O5/c1-10(2)11(12)16-9-8-15-7-6-14-5-4-13-3/h1,4-9H2,2-3H3

InChI key

OBBZSGOPJQSCNY-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service