Skip to Content
Merck
All Photos(1)

Key Documents

728098

Sigma-Aldrich

cis-Cyclopentane-1,2-dicarboxylic acid

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O4
CAS Number:
Molecular Weight:
158.15
Beilstein:
2049007
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

crystals

mp

132-136 °C

functional group

carboxylic acid

SMILES string

OC(=O)[C@H]1CCC[C@H]1C(O)=O

InChI

1S/C7H10O4/c8-6(9)4-2-1-3-5(4)7(10)11/h4-5H,1-3H2,(H,8,9)(H,10,11)/t4-,5+

InChI key

ASJCSAKCMTWGAH-SYDPRGILSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service